D-Alanine 97%

Supplier: Ambeed
Warning

Synonyms: (R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine

A109670-100G A109670-10G A109670-25G A109670-500G A109670-1000G
77093-912EA 15.92 USD
77093-912 77093-914 77093-916 77093-918 77684-985
D-Alanine 97%
D-Alanine

Precaución: for Research Use Only

Formula: C₃H₇NO₂
MW: 89.09 g/mol
MDL Number: MFCD00008077
CAS Number: 338-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR