D-(-)-Arginine 98%

Supplier: Ambeed
Warning

A109491-100G A109491-10G A109491-1G A109491-5G A109491-25G
77093-900EA 123 USD
77093-900 77093-902 77093-904 77093-906 77570-542
D-(-)-Arginine 98%
D-(-)-Arginine

Precaución: for Research Use Only

Formula: C₆H₁₄N₄O₂
MDL Number: MFCD00063116
CAS Number: 157-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR