3-Bromo-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine hydrobromide 95%

Supplier: Ambeed
Warning

A108707-100MG A108707-1G A108707-250MG
77093-828EA 30 USD
77093-828 77093-830 77093-832
3-Bromo-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine hydrobromide 95%
3-Bromo-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine hydrobromide

Precaución: for Research Use Only

Formula: C₅H₈Br₂N₄
MDL Number: MFCD22689368
CAS Number: 1414958-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR