1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A108703-10G A108703-250MG A108703-5G A108703-1G
77093-822EA 251.44 USD
77093-822 77093-824 77093-826 77684-980
1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde 97%
1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD00159623
CAS Number: 2644-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR