4-Bromo-3-chlorobenzotrifluoride 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-chloro-4-trifluoromethylbenzene

A108685-100G A108685-25G
77093-818EA 111.4 USD
77093-818 77093-820
4-Bromo-3-chlorobenzotrifluoride 98%
4-Bromo-3-chlorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
MDL Number: MFCD00673985
CAS Number: 402-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR