Z-Cha-OH 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(((Benzyloxy)carbonyl)amino)-3-cyclohexylpropanoic acid

A108538-10G A108538-1G A108538-25G A108538-5G
77093-810EA 34 USD
77093-810 77093-812 77093-814 77093-816
Z-Cha-OH 95%
Z-Cha-OH

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₄
MDL Number: MFCD00191081
CAS Number: 25341-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR