Methyl 2-(((trifluoromethyl)sulfonyl)oxy)acetate 95%

Supplier: Ambeed
Danger

A108449-250MG A108449-1G A108449-5G
77093-808EA 36.08 USD
77093-808 77684-973 77684-974
Methyl 2-(((trifluoromethyl)sulfonyl)oxy)acetate 95%
Methyl 2-(((trifluoromethyl)sulfonyl)oxy)acetate

Precaución: for Research Use Only

Formula: C₄H₅F₃O₅S
MW: 222.13 g/mol
MDL Number: MFCD23134700
CAS Number: 88016-31-3
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR