3,5-Dichloro-4-formylbenzoic acid 97%

Supplier: Ambeed
Warning

A106758-100MG A106758-1G A106758-250MG A106758-5G
77093-766EA 15.92 USD
77093-766 77093-768 77093-770 77093-772
3,5-Dichloro-4-formylbenzoic acid 97%
3,5-Dichloro-4-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄Cl₂O₃
MW: 219.02 g/mol
MDL Number: MFCD24483956
CAS Number: 153203-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR