1-Methyl-5-nitro-1H-imidazole-2-carbonitrile 96%

Supplier: Ambeed
Warning

A106411-100MG A106411-250MG
77093-758EA 12.74 USD
77093-758 77093-760
1-Methyl-5-nitro-1H-imidazole-2-carbonitrile 96%
1-Methyl-5-nitro-1H-imidazole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄N₄O₂
MW: 152.11 g/mol
MDL Number: MFCD21336309
CAS Number: 1615-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR