(2R,3R)-3-(2,5-Difluorophenyl)-3-hydroxy-2-methyl-4-(1H-1,2,4-triazol-1-yl)butanoic acid 95%

Supplier: Ambeed
Warning

A106080-5G A106080-1G A106080-250MG
77093-754EA 555.92 USD
77093-754 77684-960 77684-961
(2R,3R)-3-(2,5-Difluorophenyl)-3-hydroxy-2-methyl-4-(1H-1,2,4-triazol-1-yl)butanoic acid 95%
(2R,3R)-3-(2,5-Difluorophenyl)-3-hydroxy-2-methyl-4-(1H-1,2,4-triazol-1-yl)butanoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₃F₂N₃O₃
MW: 297.25 g/mol
MDL Number: MFCD28404610
CAS Number: 483340-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR