2-((tert-Butoxycarbonyl)amino)-3-(4-chlorophenyl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Boc-DL-Phe(4-Cl)-OH

A105710-25G A105710-5G A105710-10G
77093-740EA 8.49 USD
77093-740 77093-742 77093-744 77684-958
2-((tert-Butoxycarbonyl)amino)-3-(4-chlorophenyl)propanoic acid 98%
2-((tert-Butoxycarbonyl)amino)-3-(4-chlorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈ClNO₄
MDL Number: MFCD02081080
CAS Number: 51301-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR