2-Acetyl-5-iodothiophene 97%

Supplier: Ambeed
Danger

Synonyms: 1-(5-Iodo-2-thienyl)-1-ethanone, 1-(5-Iodothiophen-2-yl)ethanone

A105302-1G A105302-250MG A105302-5G
77093-722EA 24.41 USD
77093-722 77093-724 77093-726
2-Acetyl-5-iodothiophene 97%
2-Acetyl-5-iodothiophene

Precaución: for Research Use Only

Formula: C₆H₅IOS
MW: 252.08 g/mol
MDL Number: MFCD02093967
CAS Number: 30955-94-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR