(4,5-Dichloro-2-methylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

A105116-100MG A105116-1G A105116-250MG
77093-716EA 47 USD
77093-716 77093-718 77093-720
(4,5-Dichloro-2-methylphenyl)boronic acid 97%
(4,5-Dichloro-2-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BCl₂O₂
MDL Number: MFCD20441818
CAS Number: 1612184-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR