N-BOC-4-bromo-3-methylaniline 95%

Supplier: Ambeed
Warning

A105053-10G A105053-1G A105053-250MG A105053-5G A105053-25G
77093-708EA 120 USD
77093-708 77093-710 77093-712 77093-714 77900-755
N-BOC-4-bromo-3-methylaniline 95%
N-BOC-4-bromo-3-methylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD21170329
CAS Number: 654056-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR