5-Methylisoxazole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A104932-100G A104932-10G A104932-1G A104932-25G A104932-5G
77093-690EA 205.82 USD
77093-690 77093-692 77093-694 77093-696 77093-698
5-Methylisoxazole-3-carboxylic acid 98%
5-Methylisoxazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅NO₃
MW: 127.1 g/mol
MDL Number: MFCD01318162
CAS Number: 3405-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR