2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A104903-1G
A104903-5G
77093-688EA
40
USD
77093-688
77684-951
2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 95%
2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₅H₁₉NO₄ |
MDL Number:
MFCD02060701 CAS Number: 151838-62-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...