1-Bromo-2-chloro-3-nitrobenzene 96%

Supplier: Ambeed
Warning

A104791-10G A104791-250MG A104791-25G
77093-668EA 158.08 USD
77093-668 77093-670 77093-672
1-Bromo-2-chloro-3-nitrobenzene 96%
1-Bromo-2-chloro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MW: 236.4505 g/mol
MDL Number: MFCD08277301
CAS Number: 3970-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR