4-Chloro-7-nitroquinoline 97%

Supplier: Ambeed
Warning

A104587-100MG A104587-250MG
77093-656EA 18.04 USD
77093-656 77093-658
4-Chloro-7-nitroquinoline 97%
4-Chloro-7-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClN₂O₂
MW: 208.60 g/mol
MDL Number: MFCD08688612
CAS Number: 18436-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR