3-Chloro-5-fluorophenol 98%

Supplier: Ambeed
Warning

A104538-100G A104538-10G A104538-1G A104538-25G A104538-5G
77093-646EA 812.65 USD
77093-646 77093-648 77093-650 77093-652 77093-654
3-Chloro-5-fluorophenol 98%
3-Chloro-5-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₄ClFO
MW: 146.55 g/mol
MDL Number: MFCD00042212
CAS Number: 202982-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR