Z-εisopropyl-L-lysine 97%

Supplier: Ambeed
Warning

Synonyms: Z-Lys(isopropyl)-OH, (S)-2-(((Benzyloxy)carbonyl)amino)-6-(isopropylamino)hexanoic acid

A104018-1G A104018-250MG
77093-622EA 72.15 USD
77093-622 77093-624
Z-εisopropyl-L-lysine 97%
Z-Nepsilonisopropyl-L-lysine

Precaución: for Research Use Only

Formula: C₁₇H₂₆N₂O₄
MW: 322.405 g/mol
MDL Number: MFCD01318752
CAS Number: 218938-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR