4-Bromo-3-chloro-2-fluoroaniline 98%

Supplier: Ambeed
Warning

A102916-100G A102916-10G A102916-1G A102916-25G A102916-5G
77093-588EA 82.76 USD
77093-588 77093-590 77093-592 77093-594 77093-596
4-Bromo-3-chloro-2-fluoroaniline 98%
4-Bromo-3-chloro-2-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
MW: 224.46 g/mol
MDL Number: MFCD04039280
CAS Number: 115843-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR