2,4-Dichloro-6-isopropylpyrimidine 98%

Supplier: Ambeed
Warning

A102818-100MG A102818-250MG A102818-1G
77093-576EA 60.48 USD
77093-576 77093-578 77592-090
2,4-Dichloro-6-isopropylpyrimidine 98%
2,4-Dichloro-6-isopropylpyrimidine

Precaución: for Research Use Only

Formula: C₇H₈Cl₂N₂
MW: 191.05 g/mol
MDL Number: MFCD18916412
CAS Number: 89938-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR