Ethyl isonicotinate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Picolinic acid ethyl ester, 4-Pyridinecarboxylic acid ethyl ester, Pyridine-4-carboxylic acid ethyl ester, Ethyl 4-pyridinecarboxylate, Isonicotinic acid ethyl ester

A102100-1000G A102100-100G A102100-10G A102100-25G A102100-500G
77093-538EA 176.11 USD
77093-538 77093-540 77093-542 77093-544 77093-546
Ethyl isonicotinate 98%
Ethyl isonicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00006428
CAS Number: 1570-45-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR