4,4'-Difluorobenzophenone 98%
Supplier: Ambeed
|
Warning
|
A100862-100G
A100862-25G
A100862-500G
A100862-5G
77093-488EA
11
USD
77093-488
77093-490
77093-492
77565-754
4,4'-Difluorobenzophenone 98%
4,4'-Difluorobenzophenone
Precaución: For research use only.
Formula:
C₁₃H₈F₂O |
MDL Number:
MFCD00000353 CAS Number: 345-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...