XL-647 98%

Supplier: Ambeed
Warning

Synonyms: Tesevatinib, N-(3,4-Dichloro-2-fluorophenyl)-6-methoxy-7-(((3aR,5r,6aS)-2-methyloctahydrocyclopenta[c]pyrrol-5-yl)methoxy)quinazolin-4-amine, EXEL-7647, KD-019, N-(3,4-dichloro-2-fluorophenyl)-6-methoxy-7-(((3aR,6aS)-2-methyloctahydrocyclopenta[c]pyrrol-5-yl)methoxy)quinazolin-4-amine

A100327-10MG A100327-1MG A100327-2MG A100327-5MG
77093-458EA 304 USD
77093-458 77093-460 77093-462 77093-464
XL-647 98%
XL-647

Precaución: for Research Use Only

Formula: C₂₄H₂₅Cl₂FN₄O₂
MW: 491.38 g/mol
MDL Number: MFCD22124889
CAS Number: 781613-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR