OEM Specific Fittings
Supplier: Optimize
10-20-00330 10-20-00331 10-23-00358 40-24-01271 40-24-01273 40-24-01275 40-24-01276 40-24-01290 40-24-01291 41-24-01277 41-24-01278 41-24-01279 41-24-01280 41-24-01281 41-24-01282 10-20-02876 10-20-02875 10-20-02878 10-20-02877 41-24-01283
21510-762EA 25.99 USD
21510-762 21510-764 21510-802 77101-378 77101-380 77101-382 77101-384 77101-386 77101-388 77101-390 77101-392 77101-394 77101-396 77101-398 77101-400 83008-160 83008-158 83008-164 83008-162 89522-312
OEM Specific Fittings
Fittings and Connectors
OPTI-LOK Fittings are ideal for high-pressure connections with ¹/₁₆" OD tubing. All Optimize fittings are precision machined for optimal quality and thread consistency and are designed for trouble-free operation at pressures of up to 6,000 psi.
  • OEM Specific Fittings (Nuts, Ferrules and Unions) for ¹/₁₆" Tubing
  • Waters, Valco, Parker and SSI styles available
  • Fittings are included with all Unions
  • Precision machined for optimal quality and thread consistency
  • Zero-dead-volume connections every time
We realize that your time is valuable and should not be wasted worrying about fittings. For this reason, all Optimize fittings are precision machined in the USA for optimal quality and thread consistency. The machining process is the best way to ensure tight manufacturing tolerances and minimal product variance. What does all this mean? It means that every fitting you purchase from Optimize will perform just as well as the last, so you will have no worries about faulty fittings. This reflects our commitment to providing our customers with the highest quality, best designed product available.