Monomethyl terephthalate 98%

Supplier: Ambeed
Warning

A261585-25G A261585-1000G A261585-100G A261585-10G A261585-500G A261585-5G
77092-026EA 12.74 USD
77092-026 77262-230 77262-232 77262-234 77262-236 77262-238
Monomethyl terephthalate 98%
Monomethyl terephthalate

Precaución: for Research Use Only

Formula: C₉H₈O₄
MW: 180.16 g/mol
MDL Number: MFCD00002557
CAS Number: 1679-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR