1-Chloro-2,6-dibromo-4-fluorobenzene 97%

Supplier: Ambeed
Warning

A259593-25G A259593-100G A259593-10G A259593-5G A259593-500G
77091-902EA 31 USD
77091-902 77289-180 77289-182 77289-184 77748-489
1-Chloro-2,6-dibromo-4-fluorobenzene 97%
1-Chloro-2,6-dibromo-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂ClF
MDL Number: MFCD00061122
CAS Number: 179897-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR