9,9-Bis(4-aminophenyl)fluorene 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(9-Fluorenylidene)dianiline, 4,4'-(9H-Fluorene-9,9-diyl)dianiline

A256550-25G A256550-100G A256550-10G A256550-500G
77091-634EA 12.74 USD
77091-634 77289-138 77289-140 77289-142
9,9-Bis(4-aminophenyl)fluorene 98%
9,9-Bis(4-aminophenyl)fluorene

Precaución: for Research Use Only

Formula: C₂₅H₂₀N₂
MW: 348.45 g/mol
MDL Number: MFCD00039156
CAS Number: 15499-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR