2,2-Dimethylbutyric acid 98%

Supplier: Ambeed
Warning

A246402-25G A246402-5G A246402-100G A246402-500G
77090-838EA 15 USD
77090-838 77153-194 77612-378 77612-380
2,2-Dimethylbutyric acid 98%
2,2-Dimethylbutyric acid

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MDL Number: MFCD00004200
CAS Number: 595-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR