Diphenylmethane 99%

Supplier: Ambeed
Danger

A238651-25G A238651-100G
77090-154EA 6.37 USD
77090-154 77568-308
Diphenylmethane 99%
Diphenylmethane

Precaución: For research use only.

Formula: C₁₃H₁₂
MW: 168.24 g/mol
MDL Number: MFCD00004781
CAS Number: 101-81-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR