1,2,4-Triazole 97%

Supplier: Ambeed
Danger

Synonyms: 1,2,4-1H-Triazole, 1H-1,2,4-Triazole

A234123-25G A234123-100G A234123-500G A234123-1000G
77089-662EA 5.31 USD
77089-662 77113-088 77113-090 77568-272
1,2,4-Triazole 97%
1,2,4-Triazole

Precaución: For research use only.

Formula: C₂H₃N₃
MW: 69.07 g/mol
MDL Number: MFCD00005228
CAS Number: 288-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR