N-Acetyl-L-cysteine 98%
Supplier: Ambeed
|
Warning
|
A233740-25G
A233740-100G
A233740-10G
A233740-500G
A233740-5G
77089-640EA
11
USD
77089-640
77113-080
77113-082
77113-084
77113-086
N-Acetyl-L-cysteine 98%
N-Acetyl-L-cysteine
Precaución: for Research Use Only
Formula:
C₅H₉NO₃S |
MDL Number:
MFCD00004880 CAS Number: 616-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...