tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 2,5-dihydro-1H-pyrrole-1-carboxylate, N-Boc-2,5-dihydropyrrole, N-BOC-3-Pyrroline
A226621-25G
A226621-100G
A226621-10G
A226621-1G
A226621-500G
A226621-5G
77088-890EA
17
USD
77088-890
77112-854
77112-856
77112-858
77112-860
77568-192
tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate 97%
tert-Butyl-2,5-dihydro-1H-pyrrole-1-carboxylate
Precaución: For research use only.
Formula:
C₉H₁₅NO₂ MW: 169.224 g/mol |
MDL Number:
MFCD01863512 CAS Number: 73286-70-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...