2-Methoxynaphthalene 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-naphthyl ether

A225905-25G A225905-1000G A225905-100G A225905-500G
77088-832EA 6.37 USD
77088-832 77261-414 77261-416 77261-418
2-Methoxynaphthalene 97%
2-Methoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₁₀O
MW: 158.2 g/mol
MDL Number: MFCD00004061
CAS Number: 93-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR