Methyl nicotinate 98%

Supplier: Ambeed
Warning

Synonyms: 3-Picolinic acid methyl ester, 3-(Methoxycarbonyl)pyridine, Pyridine-3-carboxylic acid methyl ester, Nicotinic acid methyl ester, 3-Pyridinecarboxylic acid methyl ester, Methyl 3-pyridinecarboxylate

A218501-25G A218501-100G A218501-500G A218501-1000G
77088-106EA 10 USD
77088-106 77112-664 77112-666 77612-132
Methyl nicotinate 98%
Methyl nicotinate

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 209
Melting Pt: 39
Flash Pt: 95
MDL Number: MFCD00006388
CAS Number: 93-60-7
Merck Index: 12,06174

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR