(4-Fluorophenyl)-[(3S)-3-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]piperidin-1-yl]methanone 99+%

Supplier: Ambeed
Warning

Synonyms: ADX-47273, (S)-(4-Fluorophenyl)(3-(3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl)piperidin-1-yl)methanone

A217506-10MG A217506-25MG A217506-5MG A217506-1MG
77087-978EA 51.99 USD
77087-978 77087-980 77087-982 77097-864
(4-Fluorophenyl)-[(3S)-3-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]piperidin-1-yl]methanone 99+%
(4-Fluorophenyl)-[(3S)-3-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]piperidin-1-yl]methanone

Precaución: for Research Use Only

Formula: C₂₀H₁₇F₂N₃O₂
MW: 369.372 g/mol
MDL Number: MFCD17167026
CAS Number: 851881-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR