2,4-Difluorophenol 98%

Supplier: Ambeed
Danger

A207990-25G A207990-100G A207990-10G A207990-5G
77086-830EA 19 USD
77086-830 77097-608 77097-610 77097-612
2,4-Difluorophenol 98%
2,4-Difluorophenol

Precaución: for Research Use Only

Formula: C₆H₄F₂O
MDL Number: MFCD00009715
CAS Number: 367-27-1
UN: 2921
ADR: 8,4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR