Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl) 98%

Supplier: Ambeed
Warning

Synonyms: H-Gly-OtBu.HCl, Glycine tert-butyl ester hydrochloride, H-Gly-OtBu·HCl

A207246-25G A207246-10G A207246-500G A207246-100G A207246-1000G
77086-728EA 8 USD
77086-728 77275-020 77275-022 77550-330 77550-328 77615-738
Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl) 98%
Glycine-tert-butyl ester hydrochloride(H-Gly-OtBu.HCl)

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO₂
MDL Number: MFCD00058255
CAS Number: 27532-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR