4'-Aminoacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Aminophenyl)ethanone, p-Aminoacetophenone

A206379-25G A206379-1000G A206379-100G A206379-10G A206379-500G
77086-630EA 6.37 USD
77086-630 77112-332 77112-334 77112-336 77112-338
4'-Aminoacetophenone 98%
4'-Aminoacetophenone

Precaución: for Research Use Only

Formula: C₈H₉NO
MW: 135.17 g/mol
MDL Number: MFCD00007896
CAS Number: 99-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR