1-Fluoronaphthalene 98%

Supplier: Ambeed
Warning

A205033-25G A205033-100G A205033-10G A205033-500G
77086-448EA 6.37 USD
77086-448 77615-714 77615-716 77615-718
1-Fluoronaphthalene 98%
1-Fluoronaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₇F
MW: 146.16 g/mol
MDL Number: MFCD00003873
CAS Number: 321-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR