L-Arginine ethyl ester dihydrochloride 96%

Supplier: Ambeed
Warning

Synonyms: H-Arg-OEt·2 HCl, H-Arg-OEt·2HCl, H-Arg-OEt.2HCl

A198965-25G A198965-100G A198965-10G
77085-710EA 11.67 USD
77085-710 77192-222 77192-224
L-Arginine ethyl ester dihydrochloride 96%
L-Arginine ethyl ester dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₂₀Cl₂N₄O₂
MW: 275.174 g/mol
MDL Number: MFCD00038949
CAS Number: 36589-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR