4-Methyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

Synonyms: Fomepizole

A195940-25G A195940-10G A195940-1G A195940-5G A195940-100G A195940-500G
77085-358EA 29.71 USD
77085-358 77287-552 77287-554 77287-556 77748-242 77748-243
4-Methyl-1H-pyrazole 98%
4-Methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₆N₂
MW: 82.11 g/mol
MDL Number: MFCD00005245
CAS Number: 7554-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR