3-Nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A191523-25G A191523-1000G A191523-100G A191523-500G
77084-818EA 13 USD
77084-818 77151-900 77151-902 77151-904
3-Nitrobenzoic acid 98%
3-Nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅NO₄
MDL Number: MFCD00007251
CAS Number: 121-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR