Methyl-2-chloronicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloronicotinate, Methyl 2-chloropyridine-3-carboxylate

A179763-25G A179763-100G A179763-10G A179763-500G A179763-1000G
77083-284EA 9.55 USD
77083-284 77138-660 77138-662 77138-664 77746-170
Methyl-2-chloronicotinate 98%
Methyl-2-chloronicotinate

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD01318629
CAS Number: 40134-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR