4'-Phenylacetophenone 97%

Supplier: Ambeed
Warning

Synonyms: 4-Acetylbiphenyl, 1-([1,1'-Biphenyl]-4-yl)ethanone, 4-Biphenylyl methyl ketone, 4'-Phenylacetophenone, p-Phenylacetophenone, 4-Acetyl-biphenyl

A179539-25G A179539-100G A179539-10G A179539-500G A179539-5G
77083-260EA 8.49 USD
77083-260 77287-126 77287-128 77287-130 77287-132
4'-Phenylacetophenone 97%
4'-Phenylacetophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₂O
MW: 196.25 g/mol
MDL Number: MFCD00008749
CAS Number: 92-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR