5-Bromonicotinic acid 98%

Supplier: Ambeed
Warning

A177455-25G A177455-1000G A177455-100G A177455-10G A177455-500G
77082-954EA 11 USD
77082-954 77273-914 77273-916 77273-918 77273-920
5-Bromonicotinic acid 98%
5-Bromonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄BrNO₂
MDL Number: MFCD00009783
CAS Number: 20826-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR