Di-t-butylneopentylphosphonium tetrafluoroborate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Di-tert-butyl(neopentyl)phosphonium tetrafluoroborate
A999025-100MG
A999025-250MG
76994-098EA
15
USD
76994-098
76994-100
Di-t-butylneopentylphosphonium tetrafluoroborate 98%
Di-t-butylneopentylphosphonium tetrafluoroborate
Precaución: for Research Use Only
Formula:
C₁₃H₃₀BF₄P |
MDL Number:
MFCD08064039 CAS Number: 886059-84-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...