Ethyl-3-chloropicolinate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3-chloropicolinate, Ethyl 3-chloro-2-pyridinecarboxylate

A998258-1G A998258-5G
77007-308EA 22 USD
77007-308 77007-310
Ethyl-3-chloropicolinate 95%
Ethyl 3-chloropicolinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MDL Number: MFCD11656307
CAS Number: 128073-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR