2-(Benzyloxy)-5-bromobenzaldehyde 95%

Supplier: Ambeed
Warning

A997859-1G A997859-250MG A997859-5G
77037-090EA 60.48 USD
77037-090 77037-092 77037-094
2-(Benzyloxy)-5-bromobenzaldehyde 95%
2-(Benzyloxy)-5-bromobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrO₂
MW: 291.1399 g/mol
MDL Number: MFCD00046154
CAS Number: 121124-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR